Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thiobarbituric Acid Reactants (TBARS) Colorimetric Assay Kit

Catalog No. EEA021
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA021 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA021 Supplier Invitrogen™ Supplier No. EEA021
Only null left
Add to Cart
Add to Cart

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Thiobarbituric Acid Reactants (TBARS) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of TBARS in serum, plasma, animal tissue, cultured cells, and other sample types. TBARS are products of lipid peroxidation, a process that occurs when polyunsaturated fatty acids in cellular membranes are attacked by oxygen free radicals. TBARS, particularly malondialdehyde (MDA), are commonly used as biomarkers of lipid peroxidation and can reflect the degree of oxidative stress and cellular injury in organisms.

In this assay, TBARS and thiobarbituric acid (TBA) react under high temperature and acidic conditions. This reaction forms a pink compound, which has an optical density measured at 530–540 nm and the concentration of which is linearly related to the concentration of TBARS in the sample.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 2.6–100 μmol/L
Assay Sensitivity 0.85 μmol/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Plasma, Serum, Tissue Homogenates
For Use With (Equipment) Microplate Reader
Incubation Time 60 min.
Interassay CV 7.1
Intraassay CV 4.3
Quantity 96 tests
Sample Volume 100 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Thiobarbituric Acid Reactants (TBARS)
Total Assay Time 80 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.