Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Total Antioxidant Status (TAS) Colorimetric Assay Kit

Catalog No. EEA025
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA025 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA025 Supplier Invitrogen™ Supplier No. EEA025
Only null left
Add to Cart
Add to Cart

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Total Antioxidant Status (TAS) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of TAS in serum, plasma, urine, cellular supernatant, and animal and plant tissue samples. Antioxidants are substances that help protect cells against damage caused by free radicals, which are unstable molecules that can lead to oxidative stress and various diseases..

The assay uses ABTS, which is oxidized to ABTS•+ (a blue-green solution) and then further reduced back to colorless ABTS in the presence of antioxidants. The total antioxidant status can be calculated by measuring the absorbance of ABTS•+ at 660 nm.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.29–25.85 mmol/L
Assay Sensitivity 0.23 mmol Trolox Equiv./L
Content And Storage Store at –20°C.

Detection Method Colorimetric
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates, Urine
For Use With (Equipment) Microplate Reader
Incubation Time 5 min.
Interassay CV 7
Intraassay CV 4.6
Quantity 96 tests
Sample Volume 10 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Total Antioxidant Status (TAS)
Total Assay Time 30 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.