Learn More
Medchemexpress LLC Elacestrant dihydrochloride | 1349723-93-8 | 99.5% | 531.56 | 5 MG
SDP
Catalog No. 5000393334
Encompass_Preferred
Elacestrant dihydrochloride is an orally available and selective estrogen receptor degrader (SERD) that inhibits the growth of ER+ breast cancer cell lines both in vitro and in vivo. It has IC50s of 48 nM for ERα and 870 nM for ERβ. This product is for research use only and not sold to patients.
  • Orally available and selective estrogen receptor degrader
  • Inhibits the growth of ER+ breast cancer cell lines
  • For research use only
  • Appearance: white to off-white solid
  • Shipped at room temperature
  • Stored at 4°C, sealed, away from moisture
  • In solvent, stored at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-93334 Supplier Medchemexpress LLC Supplier No. 5MGHY19822A
Elacestrant dihydrochloride is an orally available and selective estrogen receptor degrader (SERD) that inhibits the growth of ER+ breast cancer cell lines both in vitro and in vivo. It has IC50s of 48 nM for ERα and 870 nM for ERβ. This product is for research use only and not sold to patients.
  • Orally available and selective estrogen receptor degrader
  • Inhibits the growth of ER+ breast cancer cell lines
  • For research use only
  • Appearance: white to off-white solid
  • Shipped at room temperature
  • Stored at 4°C, sealed, away from moisture
  • In solvent, stored at -80°C for 6 months or -20°C for 1 month