Learn More
Medchemexpress LLC Elacestrant dihydrochloride | 1349723-93-8 | 99.5% | 531.56 | 50 MG
SDP
Catalog No. 5000402148
Encompass_Preferred
Elacestrant dihydrochloride is an orally available and selective estrogen receptor degrader (SERD) that can inhibit the growth of ER+ breast cancer cell lines in vitro and in vivo. It is effective against ERα and ERβ, making it suitable for research into hormone receptor-positive breast cancers.
  • Orally available and selective estrogen receptor degrader (SERD)
  • Inhibits the growth of ER+ breast cancer cell lines in vitro and in vivo
  • Effective against ERα and ERβ targets
  • Suitable for various research applications in breast cancer studies
Catalog No. 50-004-02148 Supplier Medchemexpress LLC Supplier No. HY19822A50MG
Elacestrant dihydrochloride is an orally available and selective estrogen receptor degrader (SERD) that can inhibit the growth of ER+ breast cancer cell lines in vitro and in vivo. It is effective against ERα and ERβ, making it suitable for research into hormone receptor-positive breast cancers.
  • Orally available and selective estrogen receptor degrader (SERD)
  • Inhibits the growth of ER+ breast cancer cell lines in vitro and in vivo
  • Effective against ERα and ERβ targets
  • Suitable for various research applications in breast cancer studies