Learn More
Medchemexpress LLC Elacridar | 143664-11-3 | 99.7% | C34H33N3O5 | 1 G
SDP
Catalog No. 5000358165
Encompass_Preferred
Elacridar is an orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor. It is utilized to investigate the impact of efflux transporters on agent distribution to the brain and for cancer research.
  • Orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor
  • Used to examine the influence of efflux transporters on agent distribution to the brain
  • Useful in cancer research applications
  • Inhibits cell viability in 786-O cells
  • Affects P-glycoprotein and ABCG2 protein expression levels in MCF-7 and 786-O cell lines
  • Increases 99mTc-MIBI intracellular accumulation in MCF-7 and 786-O cells
Catalog No. 50-003-58165 Supplier Medchemexpress LLC Supplier No. HY508791G
Elacridar is an orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor. It is utilized to investigate the impact of efflux transporters on agent distribution to the brain and for cancer research.
  • Orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor
  • Used to examine the influence of efflux transporters on agent distribution to the brain
  • Useful in cancer research applications
  • Inhibits cell viability in 786-O cells
  • Affects P-glycoprotein and ABCG2 protein expression levels in MCF-7 and 786-O cell lines
  • Increases 99mTc-MIBI intracellular accumulation in MCF-7 and 786-O cells