Learn More
Selleck Chemical LLC Elacridar
SDP
Catalog No. 5000001440
Encompass_Preferred
Elacridar (GF120918 GW120918 GG918 GW0918) is a potent P-gp (MDR-1) and BCRP inhibitor
Catalog No. 50-000-01440 Supplier Selleck Chemical LLC Supplier No. S777210MM/1ML
Elacridar (GF120918 GW120918 GG918 GW0918) is a potent P-gp (MDR-1) and BCRP inhibitor