Learn More
Selleck Chemical LLC Elacridar
SDP
Supplier:  Selleck Chemical LLC S77721G
Encompass_Preferred
Elacridar (GF120918 GW120918 GG918 GW0918) is a potent P-gp (MDR-1) and BCRP inhibitor
Catalog No. 50-000-04616