Learn More
TARGETMOL CHEMICALS INC Elacridar 25MG
Supplier: TARGETMOL CHEMICALS INC T265725MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Elacridar (GG918) is a potent inhibitor of P-glycoprotein and BCRP. Purity 99.32%