Learn More
Selleck Chemical LLC Elacridar (GF120918) 50mg 143664-11-3 GW120918
SDP
Supplier:  Selleck Chemical LLC S777250MG
Encompass_Preferred
Elacridar (GF120918, GW120918) is a potent P-gp (MDR-1) and BCRP inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5231