Learn More
Medchemexpress LLC Elacridar hydrochloride | 143851-98-3 | 99.8% | C34H34ClN3O5 | 5 MG
SDP
Catalog No. 5000358871
Encompass_Preferred
Elacridar hydrochloride (GF120918A) is an orally active dual-inhibitor of P-glycoprotein (P-gp) and breast cancer resistance protein (BCRP). This compound is used for research to examine the influence of efflux transporters on agent distribution to the brain and for cancer research.
  • Orally active P-glycoprotein (P-gp) inhibitor
  • Orally active breast cancer resistance protein (BCRP) inhibitor
  • Used to examine the influence of efflux transporters on agent distribution to the brain
  • Applicable for cancer research
  • Inhibits cell viability of 786-O cells in vitro
  • Affects P-glycoprotein and ABCG2 protein expression levels in MCF-7 and 786-O cell lines
  • Increases ⁹⁹mTc-MIBI intracellular accumulation in MCF-7 and 786-O cell lines
Catalog No. 50-003-58871 Supplier Medchemexpress LLC Supplier No. HY508805MG
Elacridar hydrochloride (GF120918A) is an orally active dual-inhibitor of P-glycoprotein (P-gp) and breast cancer resistance protein (BCRP). This compound is used for research to examine the influence of efflux transporters on agent distribution to the brain and for cancer research.
  • Orally active P-glycoprotein (P-gp) inhibitor
  • Orally active breast cancer resistance protein (BCRP) inhibitor
  • Used to examine the influence of efflux transporters on agent distribution to the brain
  • Applicable for cancer research
  • Inhibits cell viability of 786-O cells in vitro
  • Affects P-glycoprotein and ABCG2 protein expression levels in MCF-7 and 786-O cell lines
  • Increases ⁹⁹mTc-MIBI intracellular accumulation in MCF-7 and 786-O cell lines