Learn More
Medchemexpress LLC Elacridar hydrochloride | 143851-98-3 | 99.9% | C34H34ClN3O5 | 100 MG
SDP
Catalog No. 5000357707
Encompass_Preferred
Elacridar hydrochloride (GF120918A) is an orally active inhibitor of P-glycoprotein and breast cancer resistance protein (BCRP). It is used to investigate the influence of efflux transporters on agent distribution to the brain and for cancer research.
  • Inhibits P-glycoprotein and BCRP
  • Used in cancer research
  • Investigates efflux transporter influence on brain distribution
Catalog No. 50-003-57707 Supplier Medchemexpress LLC Supplier No. HY50880100MG
Elacridar hydrochloride (GF120918A) is an orally active inhibitor of P-glycoprotein and breast cancer resistance protein (BCRP). It is used to investigate the influence of efflux transporters on agent distribution to the brain and for cancer research.
  • Inhibits P-glycoprotein and BCRP
  • Used in cancer research
  • Investigates efflux transporter influence on brain distribution