Learn More
Medchemexpress LLC Elacridar (Standard) | 143664-11-3 | 97.5% | C34H33N3O5 | 50 MG
SDP
Catalog No. 5000357708
Encompass_Preferred
Elacridar (Standard) is the analytical standard of Elacridar, intended for research and analytical applications. It acts as an orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor. It can be utilized to study the impact of efflux transporters on agent distribution to the brain and for cancer research, enhancing the cytotoxic effects of sunitinib and preventing multidrug resistance in renal carcinoma cells.
  • Orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor
  • Enhances cytotoxic effects and prevents multidrug resistance in cancer cells
  • Intended for research and analytical applications
Catalog No. 50-003-57708 Supplier Medchemexpress LLC Supplier No. HY50879R50MG
Elacridar (Standard) is the analytical standard of Elacridar, intended for research and analytical applications. It acts as an orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor. It can be utilized to study the impact of efflux transporters on agent distribution to the brain and for cancer research, enhancing the cytotoxic effects of sunitinib and preventing multidrug resistance in renal carcinoma cells.
  • Orally active P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor
  • Enhances cytotoxic effects and prevents multidrug resistance in cancer cells
  • Intended for research and analytical applications