Learn More
TARGETMOL CHEMICALS INC ELAFIBRANOR 25MG
SDP
Catalog No. 502576765
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Elafibranor (GFT505) is an agonist of the peroxisome proliferator-activated receptor-(alpha) (PPAR-(alpha)) and peroxisome proliferator-activated receptor-(delta) (PPAR-(delta)) with EC50 values of 45 and 175 nM respectively. purity: 99%
Catalog No. 50-257-6765 Supplier TARGETMOL CHEMICALS INC Supplier No. T440825MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Elafibranor (GFT505) is an agonist of the peroxisome proliferator-activated receptor-(alpha) (PPAR-(alpha)) and peroxisome proliferator-activated receptor-(delta) (PPAR-(delta)) with EC50 values of 45 and 175 nM respectively. purity: 99%