Learn More
Medchemexpress LLC HY-14369 5mg Medchemexpress, Elagolix sodium CAS:832720-36-2 Purity:>98%
SDP
Catalog No. 502081593
Encompass_Preferred
Medchemexpress, HY-14369 5mg Elagolix sodium CAS:832720-36-2 Elagolix sodium is a human GnRH receptor (GnRHR) antagonist with an IC50 and Ki of 0.25 and 3.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1593 Supplier Medchemexpress LLC Supplier No. HY143695MG
Medchemexpress, HY-14369 5mg Elagolix sodium CAS:832720-36-2 Elagolix sodium is a human GnRH receptor (GnRHR) antagonist with an IC50 and Ki of 0.25 and 3.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.