Learn More
Medchemexpress LLC Elaidic acid | 112-79-8 | 99.8% | 282.46 | 1 ML
SDP
Catalog No. 5000438784
Encompass_Preferred
Elaidic acid is an orally active trans fatty acid. It enhances the stemness of colorectal cancer cells by activating the Wnt/ERK1/2 pathway, promoting cell proliferation, invasion, and metastasis, and inhibiting apoptosis. This compound is utilized in research on colorectal cancer, insulin, and other related areas.
  • Inhibits the growth of *Lactobacillus*.
  • Alters *Lactobacillus* cell surface hydrophobicity.
  • Reduces basal 2-deoxyglucose uptake in muscle cells and adipocytes.
  • Promotes cell proliferation, invasion, and metastasis in colorectal cancer cells.
  • Inhibits apoptosis in colorectal cancer cells.
  • Activates the Wnt/ERK1/2 pathway.
Catalog No. 50-004-38784 Supplier Medchemexpress LLC Supplier No. HY11301610MM/1ML
Add to Cart
Add to Cart
Elaidic acid is an orally active trans fatty acid. It enhances the stemness of colorectal cancer cells by activating the Wnt/ERK1/2 pathway, promoting cell proliferation, invasion, and metastasis, and inhibiting apoptosis. This compound is utilized in research on colorectal cancer, insulin, and other related areas.
  • Inhibits the growth of *Lactobacillus*.
  • Alters *Lactobacillus* cell surface hydrophobicity.
  • Reduces basal 2-deoxyglucose uptake in muscle cells and adipocytes.
  • Promotes cell proliferation, invasion, and metastasis in colorectal cancer cells.
  • Inhibits apoptosis in colorectal cancer cells.
  • Activates the Wnt/ERK1/2 pathway.