Learn More
Cayman Chemical Elaidic Acid, Purity: ≥98%, CAS: 112-79-8, 50 mg
SDP
Catalog No. 502893081
Encompass_Preferred
A trans fatty acid; reduces arachidonic acid-induced HHT and HETE formation and increases arachidonic acid-induced synthesis of PGE2, PGF2α, PGD2, and TXB2 in isolated human platelets; induces apoptosis in HUVECs from 0.1-5 mmol/L; reduces palmitic acid-induced cardiac and hepatic autophagy in mice at 100 mg/kg
Catalog No. 50-289-3081 Supplier Cayman Chemical Supplier No. 9025050 MG
A trans fatty acid; reduces arachidonic acid-induced HHT and HETE formation and increases arachidonic acid-induced synthesis of PGE2, PGF2α, PGD2, and TXB2 in isolated human platelets; induces apoptosis in HUVECs from 0.1-5 mmol/L; reduces palmitic acid-induced cardiac and hepatic autophagy in mice at 100 mg/kg