Learn More
Cayman Chemical ElaIdIc AcId-d17 10mg
SDP
Catalog No. 502938902
Encompass_Preferred
An internal standard for the quantification of elaidic acid by GC- or LC-MS
Catalog No. 50-293-8902 Supplier Cayman Chemical Supplier No. 2771510 MG
An internal standard for the quantification of elaidic acid by GC- or LC-MS