Learn More
Medchemexpress LLC Elaidic acid-d9 | 1809227-37-9 | 99.0% | 291.52 | 1 MG
SDP
Catalog No. 5000439793
Encompass_Preferred
Elaidic acid-d9 is deuterium labeled Elaidic acid. Elaidic acid is the major trans fat found in hydrogenated vegetable oils and can be used as a pharmaceutical solvent. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-39793 Supplier Medchemexpress LLC Supplier No. HY113016S1MG
Add to Cart
Add to Cart
Elaidic acid-d9 is deuterium labeled Elaidic acid. Elaidic acid is the major trans fat found in hydrogenated vegetable oils and can be used as a pharmaceutical solvent. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.