Learn More
Medchemexpress LLC Elaidic acid-d9 | 1809227-37-9 | 99.0% | 291.52 g·mol⁻1 | C18H25D9O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY113016S5MG
Encompass_Preferred
Elaidic acid-d9 is a deuterium-labeled form of elaidic acid designed for analytical and tracer applications. It serves as an internal standard for quantitative measurements and is supplied at high purity for use in small-scale laboratory analyses. The material is suitable for mass spectrometry and NMR workflows and is typically provided in milligram quantities.
  • Deuterium-labeled tracer and internal standard for quantitative analysis.
  • High purity suitable for analytical workflows (99.0%).
  • Compatible with NMR, GC-MS, and LC-MS methods.
  • Available in small quantities for lab-scale experiments.
  • Stable isotope labeling facilitates accurate quantitation and tracer studies.
Catalog No. 50-002-62419