Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Elaiophylin | 37318-06-2 | 98.0% | 1025.27 g/mol | C54H88O18 | 10 MG
SDP

Catalog No. 5000286351
Encompass_Preferred

Elaiophylin is a macrolide antibiotic and autophagy inhibitor commonly used as a research reagent. It exhibits antitumor activity in multiple cancer cell lines and is employed in cellular assays to study autophagy, cytotoxicity, and related mechanisms. The compound is supplied as a solid powder appropriate for laboratory use.

  • Macrolide antibiotic and autophagy inhibitor.
  • Reported antitumor activity in multiple cancer cell lines.
  • High purity (98.0%).
  • Chemical formula C54H88O18; molecular weight 1025.27 g/mol; CAS 37318-06-2.
  • Solid white to off-white powder suitable for cell-based assays.
  • Storage: powder stable at -20°C for about 3 years; in solvent: -80°C for 6 months or -20°C for 1 month.
  • Pack size suitable for research applications (10 mg typical).

Catalog No. 50-002-86351 Supplier Medchemexpress LLC Supplier No. HY1518410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Elaiophylin is a macrolide antibiotic and autophagy inhibitor commonly used as a research reagent. It exhibits antitumor activity in multiple cancer cell lines and is employed in cellular assays to study autophagy, cytotoxicity, and related mechanisms. The compound is supplied as a solid powder appropriate for laboratory use.

  • Macrolide antibiotic and autophagy inhibitor.
  • Reported antitumor activity in multiple cancer cell lines.
  • High purity (98.0%).
  • Chemical formula C54H88O18; molecular weight 1025.27 g/mol; CAS 37318-06-2.
  • Solid white to off-white powder suitable for cell-based assays.
  • Storage: powder stable at -20°C for about 3 years; in solvent: -80°C for 6 months or -20°C for 1 month.
  • Pack size suitable for research applications (10 mg typical).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.