Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Elaiophylin | 37318-06-2 | C54H88O18 | 5 MG
SDP

Catalog No. 5000373584
Encompass_Preferred

Elaiophylin is an autophagy inhibitor that exhibits antitumor activity in ovarian cancer cells. With a molecular weight of 1025.27, it appears as a white to off-white solid and is intended for research use only.

  • Autophagy inhibitor
  • Exhibits antitumor activity in ovarian cancer cells
  • Molecular weight of 1025.27
  • White to off-white solid appearance
  • Soluble in DMSO for in vitro experiments
  • Affects ovarian cancer cell survival during hypoxia
  • Leads to activation of cleaved CASP9/caspase-9 and PARP1
  • Downregulates BIRC5/survivin
  • Suppresses ovarian cancer SKOV3 cell growth in athymic mice
  • Achieves significant antitumor effects without toxicity at lower doses

Catalog No. 50-003-73584 Supplier Medchemexpress LLC Supplier No. HY151845MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Elaiophylin is an autophagy inhibitor that exhibits antitumor activity in ovarian cancer cells. With a molecular weight of 1025.27, it appears as a white to off-white solid and is intended for research use only.

  • Autophagy inhibitor
  • Exhibits antitumor activity in ovarian cancer cells
  • Molecular weight of 1025.27
  • White to off-white solid appearance
  • Soluble in DMSO for in vitro experiments
  • Affects ovarian cancer cell survival during hypoxia
  • Leads to activation of cleaved CASP9/caspase-9 and PARP1
  • Downregulates BIRC5/survivin
  • Suppresses ovarian cancer SKOV3 cell growth in athymic mice
  • Achieves significant antitumor effects without toxicity at lower doses

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.