Learn More
AdipoGen Elaiophylin
SDP
Supplier:  AdipoGen BVT0185M001
Encompass_Preferred
Chemical. CAS 37318-06-2. Formula C54H88O18. MW 1025.3. Isolated from Streptomyces sp. MT-1. Antibiotic. Active against Gram-positive bacteria, protozoa and tumors. Testosterone 5alpha-reductase inhibitor. Nitric oxide synthase NOS inhibitor. Immunosuppressive. Enhances the antifungal activity of rapamycin.
Catalog No. 50-256-2077