Learn More
Selleck Chemical LLC Elamipretide5mg
SDP
Supplier:  Selleck Chemical LLC S98035MG
Encompass_Preferred
Elamipretide (MTP-131, SS-31, D-Arg-Dmt-Lys-Phe-NH2) is a cytochrome c peroxidase inhibitor. Elamipretide improves mitochondrial dysfunction, synaptic and memory impairment induced by lipopolysaccharide in mice. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3509