Learn More
TARGETMOL CHEMICALS INC ELAROFIBAN 5MG
SDP
Catalog No. 502580634
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Elarofiban(RWJ-53308) is a novel and orally active GPIIb/IIIa antagonist. purity: 99%
Catalog No. 50-258-0634 Supplier TARGETMOL CHEMICALS INC Supplier No. T272505MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Elarofiban(RWJ-53308) is a novel and orally active GPIIb/IIIa antagonist. purity: 99%