Learn More
TARGETMOL CHEMICALS INC Elarofiban TFA 1MG
SDP
Catalog No. 502878215
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Elarofiban TFA (RWJ-53308 TFA) is a novel and orally active and selective GPIIb/IIIa antagonist for the study of cardiovascular disease. Purity 100%
Catalog No. 50-287-8215 Supplier TARGETMOL CHEMICALS INC Supplier No. T27250L1MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Elarofiban TFA (RWJ-53308 TFA) is a novel and orally active and selective GPIIb/IIIa antagonist for the study of cardiovascular disease. Purity 100%