Learn More
AdipoGen Elasnin
SDP
Supplier:  AdipoGen BVT0342M001
Encompass_Preferred
Chemical. CAS 68112-21-0. Formula C24H40O4. MW 392.6. Isolated from Streptomyces sp. Inhibitor of human leukocyte elastase IC50 = 1.3 µg/ml, different other elastases and chymotrypsin IC50 = 82 µg/ml.
Catalog No. 50-256-1980