Learn More
Worthington Biochemical Corporation ELASTASE 100MG
SDP
Catalog No. NC9162307
Encompass
Two times crystallized. Supplied as an aqueous suspension. This preparation must be diluted to dissolve the enzyme. The diluted enzyme should be 0.22 micron filtered before use. Suitable for the isolation of Type II lung cells.
Catalog No. NC9162307 Supplier Worthington Biochemical Corporation Supplier No. LS002279
Two times crystallized. Supplied as an aqueous suspension. This preparation must be diluted to dissolve the enzyme. The diluted enzyme should be 0.22 micron filtered before use. Suitable for the isolation of Type II lung cells.