Learn More
Worthington Biochemical Corporation ELASTASE 25 MG
SDP
Catalog No. NC9329382
Encompass
Elastase, Suspension, Code: ES, 2X Crystallized, Supplied as an aqueous suspension, Suitable for the Isolation of Type II lung cells, Store at 2 deg. to 8 deg.C, The diluted enzyme should be 0.22u filtered before use, 25mg
Catalog No. NC9329382 Supplier Worthington Biochemical Corporation Supplier No. LS002274
Elastase, Suspension, Code: ES, 2X Crystallized, Supplied as an aqueous suspension, Suitable for the Isolation of Type II lung cells, Store at 2 deg. to 8 deg.C, The diluted enzyme should be 0.22u filtered before use, 25mg