Learn More
TRIBIOSCIENCE INC Elastase, 5MG
SDP
Catalog No. NC4034451
Encompass
Product Details: Form: Freeze-dried powder; Solubility: Distilled water or dilute buffer; Stability: Store at -20° C (-4° F); Activity: 10 U/mg protein; Protein: 90%
Catalog No. NC4034451 Supplier TRIBIOSCIENCE INC Supplier No. TBP00675MG
Product Details: Form: Freeze-dried powder; Solubility: Distilled water or dilute buffer; Stability: Store at -20° C (-4° F); Activity: 10 U/mg protein; Protein: 90%