Learn More
Athens Research And Technology Inc Elastase, Native Human Neutrophils, ≥95% (SDS-PAGE), 100 µg
Catalog No. 50489731
Encompass_Preferred
Elastase is a native protein purified from human neutrophils with ≥95% (SDS-PAGE), supplied lyophilized in a 100 µg pack. Molecular weight: 29.5 kDa. Gene/protein symbol: ELANE. Elastase, Human Neutrophil: 100 UG; Salt-Free lyophilized solid.; MW 29,500; EC = 0.985; Activity: 20-22 units per mg protein; Purity: > =95% by SDS-PAGE. Common research applications include protease activity, enzyme characterization and biochemical studies.
Catalog No. 50-489-731 Supplier Athens Research And Technology Inc Supplier No. 1614051200100UG
Elastase is a native protein purified from human neutrophils with ≥95% (SDS-PAGE), supplied lyophilized in a 100 µg pack. Molecular weight: 29.5 kDa. Gene/protein symbol: ELANE. Elastase, Human Neutrophil: 100 UG; Salt-Free lyophilized solid.; MW 29,500; EC = 0.985; Activity: 20-22 units per mg protein; Purity: > =95% by SDS-PAGE. Common research applications include protease activity, enzyme characterization and biochemical studies.