Learn More
Athens Research And Technology Inc ELASTASE HU NEUTROPHIL 1MG
Catalog No. 50489733
Encompass_Preferred
Elastase, Human Neutrophil: 1 MG; Salt-Free lyophilized solid.; MW 29,500; EC = 0.985; Activity: 20-22 units per mg protein; Purity: > =95% by SDS-PAGE. This native protein is prepared and purified from whole blood in the Athens Research & Technology laboratory. Molecular Biology Reagent
Catalog No. 50-489-733 Supplier Athens Research And Technology Inc Supplier No. 16140512001MG
Elastase, Human Neutrophil: 1 MG; Salt-Free lyophilized solid.; MW 29,500; EC = 0.985; Activity: 20-22 units per mg protein; Purity: > =95% by SDS-PAGE. This native protein is prepared and purified from whole blood in the Athens Research & Technology laboratory. Molecular Biology Reagent