Learn More
Description
Specifications
Specifications
| Formulation | Salt-free. |
| Preparation Method | Reconstitute in 50mM sodium acetate, pH 5.5, with 200mM NaCl or in other aqueous buffers. Material is most active near a neutral pH, but to maintain stability upon reconstitution, use in a slightly acidic buffer (pH 5.5) to avoid autolysis. |
| Purification Method | Major band by SDS-PAGE |
| Quantity | 100 μg |
| Immunogen | Elastase |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C) for long term storage or refrigerate (4°C) for short-term storage. Acidic stock solutions are stable for up to 1 month at 4°C or for up to 1 year at −20°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.