Learn More
Worthington Biochemical Corporation Elastase, Lyophilized, 100mg
SDP
Catalog No. NC9030467
Encompass
Two times crystallized, (Code: ESL), supplied as a dialyzed, lyophilized powder. The enzyme should be 0.22 micron filtered after reconstitution and prior to use. Suitable for the isolation of Type II lung cells. Store at 2-8°C. Porcine pancreatic elastase has a molecular weight of 25.9 kDa, and a pH optimum of 8.5. While elastase will hydrolyze a wide variety of protein substrates, it is unique among proteases in its ability to hydrolyze native elastin, a substrate not attacked by trypsin, chymotrypsin or pepsin. Soybean trypsin inhibitor and kallikrein inhibitor suppress proteolytic but not elastolytic activity.
Catalog No. NC9030467 Supplier Worthington Biochemical Corporation Supplier No. LS002292
Two times crystallized, (Code: ESL), supplied as a dialyzed, lyophilized powder. The enzyme should be 0.22 micron filtered after reconstitution and prior to use. Suitable for the isolation of Type II lung cells. Store at 2-8°C. Porcine pancreatic elastase has a molecular weight of 25.9 kDa, and a pH optimum of 8.5. While elastase will hydrolyze a wide variety of protein substrates, it is unique among proteases in its ability to hydrolyze native elastin, a substrate not attacked by trypsin, chymotrypsin or pepsin. Soybean trypsin inhibitor and kallikrein inhibitor suppress proteolytic but not elastolytic activity.