Learn More
Worthington Biochemical Corporation ELASTASE LYOPHILIZED 1GM
SDP
Catalog No. 50592321
Encompass
Two times crystallized, (Code: ESL), supplied as a dialyzed, lyophilized powder. The enzyme should be 0.22 micron filtered after reconstitution and prior to use. Suitable for the isolation of Type II lung cells. 2594-83-7
Catalog No. 50-592-321 Supplier Worthington Biochemical Corporation Supplier No. LS002294
Two times crystallized, (Code: ESL), supplied as a dialyzed, lyophilized powder. The enzyme should be 0.22 micron filtered after reconstitution and prior to use. Suitable for the isolation of Type II lung cells. 2594-83-7