Learn More
Medchemexpress LLC Elastase, Porcine pancreas | 39445-21-1 | 25898.13 | 10 MG
SDP
Catalog No. 5000399617
Encompass_Preferred
Elastase, Porcine pancreas is a serine protease derived from pig pancreas, composed of a single polypeptide chain of 240 amino acid residues. It functions by hydrolyzing proteins and polypeptides, and research indicates it can induce emphysema in hamsters.
  • Derived from pig pancreas
  • Serine protease
  • Hydrolyzes proteins and polypeptides
  • Suitable for research applications only
  • Appears as a solid, white to off-white substance
  • Store at -20°C for up to 2 years
  • Ships at room temperature (within continental US)
Catalog No. 50-003-99617 Supplier Medchemexpress LLC Supplier No. HYP297410MG
Elastase, Porcine pancreas is a serine protease derived from pig pancreas, composed of a single polypeptide chain of 240 amino acid residues. It functions by hydrolyzing proteins and polypeptides, and research indicates it can induce emphysema in hamsters.
  • Derived from pig pancreas
  • Serine protease
  • Hydrolyzes proteins and polypeptides
  • Suitable for research applications only
  • Appears as a solid, white to off-white substance
  • Store at -20°C for up to 2 years
  • Ships at room temperature (within continental US)