Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Elastase, porcine pancreas aqueous suspension

Catalog No. AAJ61753MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61753MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61753MC Supplier Thermo Scientific Chemicals Supplier No. J61753MC
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 39445-21-125 kd g/mol

Elastase is used in combination with other proteases for protein analysis by mass spectrometry. Elastase, is used in tissue dissociation because elastin is found in highest concentrations in the elastic fibers of connective tissues, elastase is frequently used to dissociate tissues that contain extensive intercellular fiber networks. It is usually used with other enzymes such as collagenase, trypsin, and chymotrypsin, membrane protein solubilization, protein sequence studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A serine protease that acts on elastin
  • Suitable for the isolation of Type II lung cells
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39445-21-1
Synonym E.C. 3.4.21.36; Pancreatopeptidase E

Specifications

For Use With (Application) A serine protease that acts on elastin. Suitable for the isolation of Type II lung cells
Molecular Weight (g/mol) 25 kd
Content And Storage 2°C to 8°C
Form Suspension,MW 25000
Quantity 100 mg
Product Type Elastase

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.