Learn More
Abcam Elastase Substrate, Fluorogenic (MeOSuc-AAPV-AFC), 5MG
Catalog No. 502215320
Encompass_Preferred
MW 681.67 g/mol, Purity >99%. Highly sensitive coumarin-fluorogenic substrate for neutrophil elastase (Km =130 μM for leukocyte elastase). Compatible with in vitro assays with purified enzyme, on live cells, or in conditioned medium. The excitation wavelength is 380 nm and the emission wavelength is 500 nm.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5320 Supplier Abcam Supplier No. AB1421785MG
MW 681.67 g/mol, Purity >99%. Highly sensitive coumarin-fluorogenic substrate for neutrophil elastase (Km =130 μM for leukocyte elastase). Compatible with in vitro assays with purified enzyme, on live cells, or in conditioned medium. The excitation wavelength is 380 nm and the emission wavelength is 500 nm.
The product is subject to the following: Abcam Restricted Use Statement