Learn More
Worthington Biochemical Corporation ELASTASE SUSPENSION 1GM
SDP
Catalog No. 50592320
Encompass
Two times crystallized. Supplied as an aqueous suspension. This preparation must be diluted to dissolve the enzyme. The diluted enzyme should be 0.22 micron filtered before use. Suitable for the isolation of Type II lung cells. 2594-83-7
Catalog No. 50-592-320 Supplier Worthington Biochemical Corporation Supplier No. LS002280
Two times crystallized. Supplied as an aqueous suspension. This preparation must be diluted to dissolve the enzyme. The diluted enzyme should be 0.22 micron filtered before use. Suitable for the isolation of Type II lung cells. 2594-83-7