Learn More
Cayman Chemical ElastatInal 10mg
SDP
Catalog No. 502987653
Encompass_Preferred
A potent inhibitor of pancreatic elastase (Ki = 240 nM); more strongly inhibits pancreatic elastase versus the leukocyte derived enzyme; reduces electrophoretic mobilities of 2A proteinases from poliovirus and human rhinovirus 14 in a dose-dependent manner; inhibits elastase-like protease collagenolytic protease from C. maenas digestive glands
Catalog No. 50-298-7653 Supplier Cayman Chemical Supplier No. 2228810 MG
A potent inhibitor of pancreatic elastase (Ki = 240 nM); more strongly inhibits pancreatic elastase versus the leukocyte derived enzyme; reduces electrophoretic mobilities of 2A proteinases from poliovirus and human rhinovirus 14 in a dose-dependent manner; inhibits elastase-like protease collagenolytic protease from C. maenas digestive glands