Learn More
Diagnostic Biosystems ELASTIC Elastic
Catalog No. 502407317
Encompass_Preferred
The Elastic Stain Kit (Modified Verhoff’s) is intended for use in histological demonstration of elastin in tissue sections. Demonstration of elastic tissue is useful in cases of emphysema (atrophy of elastic tissue), arteriosclerosis (thinning and loss of elastic fibers) and various other vascular diseases.
Catalog No. 50-240-7317 Supplier Diagnostic Biosystems Supplier No. KT012
The Elastic Stain Kit (Modified Verhoff’s) is intended for use in histological demonstration of elastin in tissue sections. Demonstration of elastic tissue is useful in cases of emphysema (atrophy of elastic tissue), arteriosclerosis (thinning and loss of elastic fibers) and various other vascular diseases.