Learn More
OHAUS Elastic Bands 2
Catalog No. 01804387 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-387 Shaker Attachment
1 options
Catalog No. 01-804-387 Supplier OHAUS Supplier No. 83041434
Add to Cart
Add to Cart
Two Elastic Bands for use with Recessed Platform 504-0038-00.
  • For use with the Vortex Genie 2 family of mixers and the Vortex Genie Pulse.
  • Sample vessels of up to 500 mL can be used as well as microplates.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment