Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elastic Stain Kit, 1 Kit, G-Biosciences
SCI_ED

Catalog No. S001112
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001112 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001112 Supplier G-Biosciences Supplier No. 7862237
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Elastic Stain kit is used for histologic staining of elastin in tissue sections.

Elastin is protein occurring either as branching fibers or sheets. It is highly refractive, elastic and usually thinner than collagen fibers. Demonstration of elastic tissue is useful in various vascular diseases such as emphysema (atrophy of elastic tissue) and arteriosclerosis (thinning and loss of elastic fibers).

  • Kit components: Hematoxylin
  • Solution: 250 mL
  • Ferric Chloride Solution: 125 mL
  • Iodine Solution: 250 mL
  • Van Gieson Solution: 250 mL

Specifications

Quantity 1 Kit
Product Type Elastic Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.