Learn More
Scytek Laboratories Elastic Stain Kit (Modified Verhoff's): Large
SDP
Catalog No. NC1969838
Encompass_Preferred
The Elastic Stain Kit is intended for use in histological demonstration of elastin in tissue sections. Demonstration of elastic tissue is useful in cases of emphysema (atrophy of elastic tissue), arteriosclerosis (thinning and loss of elastic fibers) and various other vascular diseases.Elastic fibers: Black to Blue/BlackNuclei: Blue to BlackCollagen: RedMuscle & Other: YellowKit contents: HSV250, Hematoxylin Solution (5%), 250ml.FCC125, Ferric Chloride (10%, Aqueous), 125 ml.LIS125, Lugol’s Iodine Solution, 125 ml.FCB125, Ferric Chloride (2%) Differentiating Solution, 125 ml.STB125, Sodium Thiosulfate Solution (5%), 125 ml.VGS125, Van Gieson’s Solution, 125 ml.
This item is not returnable. View return policy
Catalog No. NC1969838 Supplier Scytek Laboratories Supplier No. ETS1
The Elastic Stain Kit is intended for use in histological demonstration of elastin in tissue sections. Demonstration of elastic tissue is useful in cases of emphysema (atrophy of elastic tissue), arteriosclerosis (thinning and loss of elastic fibers) and various other vascular diseases.Elastic fibers: Black to Blue/BlackNuclei: Blue to BlackCollagen: RedMuscle & Other: YellowKit contents: HSV250, Hematoxylin Solution (5%), 250ml.FCC125, Ferric Chloride (10%, Aqueous), 125 ml.LIS125, Lugol’s Iodine Solution, 125 ml.FCB125, Ferric Chloride (2%) Differentiating Solution, 125 ml.STB125, Sodium Thiosulfate Solution (5%), 125 ml.VGS125, Van Gieson’s Solution, 125 ml.