Learn More
Scytek Laboratories Elastic Stain Kit (Modified Verhoff's): Small
SDP
Catalog No. NC1969839
Encompass_Preferred
The Elastic Stain Kit is intended for use in histological demonstration of elastin in tissue sections. Demonstration of elastic tissue is useful in cases of emphysema (atrophy of elastic tissue), arteriosclerosis (thinning and loss of elastic fibers) and various other vascular diseases.Elastic fibers: Black to Blue/BlackNuclei: Blue to BlackCollagen: RedMuscle & Other: YellowKit contents: HSV060, Hematoxylin Solution (5%), 60ml.FCC030, Ferric Chloride (10%, Aqueous), 30 ml.LIS030, Lugol’s Iodine Solution, 30 ml.FCB060, Ferric Chloride (2%) Differentiating Solution, 60 ml.STB030, Sodium Thiosulfate Solution (5%), 30 ml.VGS030, Van Gieson’s Solution, 30 ml.
This item is not returnable. View return policy
Catalog No. NC1969839 Supplier Scytek Laboratories Supplier No. ETS2
The Elastic Stain Kit is intended for use in histological demonstration of elastin in tissue sections. Demonstration of elastic tissue is useful in cases of emphysema (atrophy of elastic tissue), arteriosclerosis (thinning and loss of elastic fibers) and various other vascular diseases.Elastic fibers: Black to Blue/BlackNuclei: Blue to BlackCollagen: RedMuscle & Other: YellowKit contents: HSV060, Hematoxylin Solution (5%), 60ml.FCC030, Ferric Chloride (10%, Aqueous), 30 ml.LIS030, Lugol’s Iodine Solution, 30 ml.FCB060, Ferric Chloride (2%) Differentiating Solution, 60 ml.STB030, Sodium Thiosulfate Solution (5%), 30 ml.VGS030, Van Gieson’s Solution, 30 ml.