Learn More
Sigma-Aldrich Elastic TISSUE-TROL™ Control Slides
Catalog No. 01258880 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25/Pk.
Catalog No. Quantity
01-258-880 25/Pk.
1 options
Catalog No. 01-258-880 Supplier Sigma-Aldrich Supplier No. TTR00325EA
Add to Cart
Add to Cart
Human skin containing elastic fibers
Box contains 24 unstained slides plus one reference slide stained using Elastic Stain Procedure No. HT25.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Control Slide
Quantity 25/Pk.