Learn More
Newcomer Supply Elastic, Verhoeff Stain Kit, 500 ml kit
SDP
Catalog No. NC2102269
Encompass
Newcomer Supply Elastic, Verhoeff Stain Kit procedure, commonly referred to as Verhoeff-Van Gieson (VVG) technique, is used to demonstrate pathologic changes in elastic fibers as well as demonstration of normal elastic tissue such as arteries and veins.
This item is not returnable. View return policy
Catalog No. NC2102269 Supplier Newcomer Supply Supplier No. 9116B
Newcomer Supply Elastic, Verhoeff Stain Kit procedure, commonly referred to as Verhoeff-Van Gieson (VVG) technique, is used to demonstrate pathologic changes in elastic fibers as well as demonstration of normal elastic tissue such as arteries and veins.