Learn More
Diagnostic Biosystems ELASTIN Elastin
Catalog No. 502407326
Encompass_Preferred
This antibody recognizes insoluble elastin, α-elastin, soluble non-cross linked precursor of elastin (tropoelastin). Elastin is an important polymeric protein of connective tissue that imparts elasticity to vertebrate elastic tissues.
Catalog No. 50-240-7326 Supplier Diagnostic Biosystems Supplier No. PDM300
This antibody recognizes insoluble elastin, α-elastin, soluble non-cross linked precursor of elastin (tropoelastin). Elastin is an important polymeric protein of connective tissue that imparts elasticity to vertebrate elastic tissues.