Learn More
MOLECULAR DEPOT LLC Elastin Congo Red -100 mg
Catalog No. 5000013516
Encompass_Preferred
Elastin Conjugated to Congo Red is a powdered Elastin Congo red substrate-dye conjugate used to probe elastase activity This product has been used as molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only
Catalog No. 50-000-13516 Supplier MOLECULAR DEPOT LLC Supplier No. B2014125
Elastin Conjugated to Congo Red is a powdered Elastin Congo red substrate-dye conjugate used to probe elastase activity This product has been used as molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only