Learn More
Bioss ELAVL1 Recombinant Monoclonal
SDP
Catalog No. 5000065778
Encompass_Preferred
ELAVL1 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IHC-P IHC-F IF(ICC) IP IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-65778 Supplier Bioss Supplier No. BSM61688R
ELAVL1 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IHC-P IHC-F IF(ICC) IP IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol