Learn More
Aviva Systems Biology Corp ELAVL4 Antibody - N-terminal region (ARP40971_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40971_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELAVL4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ELAVL4. Alternate names: HUD, PNEMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57680